Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081397.1 | NP_001074866.1 | 25, 26 | 3331 | ||
GenBank mRNA | AB429291.1 | - | 22, 23 | 2837 | ||
AK034257.1 | - | 2, 3 | 205 | |||
AK082350.1 | - | 25 | 3067 | |||
AK220356.1 | - | 6, 7 | 763 | |||
BC082311.1 | - | 25, 26 | 3197 | |||
BC151049.1 | - | 25, 26 | 3285 | 9087962 | ||
BC151051.1 | - | 25, 26 | 3286 | 9087964 | ||
BC157966.1 | - | 25, 26 | 3286 | 9007290 | ||
BC172122.1 | - | 25, 26 | 3286 | 9054408 |