Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081149.1 | NP_001074618.1 | 13 | 2475 | ||
GenBank mRNA | AK028058.1 | - | 12 | 2394 | ||
AK036885.1 | - | 13 | 2470 | |||
AK082463.1 | - | 13 | 2478 | |||
AK132474.1 | - | 12 | 2383 | |||
AK143408.1 | - | 13 | 2466 | |||
BC024786.1 | - | 5 | 601 |