Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sprtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111141.1 | NP_001104611.1 | 4 | 898 | ||
GenBank mRNA | AK154380.1 | - | 4 | 901 | ||
BC158088.1 | - | 4 | 753 | 9007378 |