Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sipa1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081337.1 | NP_001074806.1 | 2 | 1701 | ||
GenBank mRNA | AK028440.1 | - | 2 | 815 | ||
BC072593.1 | - | 4 | 1987 | |||
BC150682.1 | - | 2 | 1746 | 9087593 |