Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033359.2 | NP_001028531.2 | 21 | 3363 | ||
NM_001170787.1 | NP_001164258.1 | 25 | 4027 | |||
GenBank mRNA | AK138164.1 | - | 21 | 3364 | ||
AK162856.1 | - | 21 | 3363 |