Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081152.1 | NP_001074621.1 | 18 | 4528 | ||
GenBank mRNA | AK042624.1 | - | 2 | 1201 | ||
AK053074.1 | - | 2 | 1177 | |||
AK080294.1 | - | 18 | 4536 | |||
BC024717.1 | - | 2 | 550 |