Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dkc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030307.2 | NP_001025478.1 | 15 | 2522 | ||
GenBank mRNA | AK136418.1 | - | 15 | 2308 | ||
AK166105.1 | - | 15 | 2298 | |||
AK167973.1 | - | 15 | 2273 | |||
BC068171.1 | - | 15 | 2278 | |||
BC099966.1 | - | 15 | 2362 | 6435181 | ||
CF751662.1 | - | 1 | 280 |