Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh11x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271809.1 | NP_001258738.1 | 1, 2 | 319 | ||
NM_001271810.1 | NP_001258739.1 | 1, 2 | 319 | |||
GenBank mRNA | AJ564976.1 | - | 1, 2 | 386 | ||
AK039071.1 | - | 1, 2 | 320 | |||
BC126930.1 | - | 1, 2 | 242 | 40086368 |