Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081499.2 | NP_001074968.1 | 5 | 952 | ||
GenBank mRNA | BC147581.1 | - | 5 | 815 | 9056571 | |
BC151057.1 | - | 5 | 815 | 9087970 |