Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017270.2 | NP_058966.2 | 8, 9 | 1154 | ||
GenBank mRNA | BC127504.1 | - | 8, 9 | 1154 | 7325543 | |
X90710.1 | - | 8, 9 | 1180 |