Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081429.1 | NP_001074898.1 | 6, 7 | 1111 | ||
GenBank mRNA | AK035850.1 | - | 6, 7 | 814 | ||
AK041803.1 | - | 6, 7 | 870 | |||
AK045055.1 | - | 6, 7 | 1100 | |||
AK133297.1 | - | 6, 7 | 1077 | |||
AK138231.1 | - | 6, 7 | 1104 | |||
AK164955.1 | - | 6, 7 | 1078 |