Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139224.1 | NP_631970.1 | 2 | 211 | ||
GenBank mRNA | AJ437650.1 | - | 2 | 211 |