Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044382.1 | NP_001037847.1 | 3 | 393 | ||
GenBank mRNA | AF487281.1 | - | 2 | 263 | ||
AK044344.1 | - | 2 | 312 | |||
BC042707.1 | - | 3 | 308 | 4975079 | ||
BC117006.1 | - | 3 | 393 | 40126325 | ||
BC117008.1 | - | 3 | 393 | 40126327 |