Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017277.1 | NP_058973.1 | 2 | 140 | ||
GenBank mRNA | M77245.1 | - | 2 | 140 |