Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1036 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207142.2 | NP_997025.1 | 1 | 189 | ||
GenBank mRNA | BC145856.1 | - | 1 | 189 | 40131159 |