Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004075.4 | NP_004066.1 | 1, 2 | 1056 | ||
GenBank mRNA | AK290552.1 | - | 1, 2 | 356 | ||
BC017466.1 | - | 1, 2 | 479 | |||
BC030519.1 | - | 1, 2 | 729 | 5169081 | ||
D83702.1 | - | 1, 2 | 746 | |||
D84657.1 | - | 1, 2 | 160 |