Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brpf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081315.1 | NP_001074784.1 | 13 | 3779 | ||
GenBank mRNA | AK129322.1 | - | 10 | 2626 | ||
AK147555.1 | - | 13 | 3778 | |||
AK162638.1 | - | 6 | 627 | |||
BC048580.1 | - | 5 | 558 | |||
BC157915.1 | - | 12 | 3544 | 9088158 | ||
BC157957.1 | - | 11 | 3446 | |||
BC172121.1 | - | 9 | 3269 |