Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Znf512b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164597.1 | NP_001158069.1 | 17 | 3405 | ||
GenBank mRNA | AK129308.1 | - | 17 | 3405 | ||
AK159002.1 | - | 2 | 1095 | |||
BC056460.1 | - | 15 | 3392 |