Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081323.2 | NP_001074792.1 | 22 | 3829 | ||
NM_001277867.1 | NP_001264796.1 | 23 | 3919 | |||
GenBank mRNA | AK016065.1 | - | 4 | 730 | ||
BC076624.1 | - | 22 | 3692 | |||
BC138423.1 | - | 19 | 3231 | 8861443 | ||
BC145681.1 | - | 19 | 3231 | 40130984 |