Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Megf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160400.1 | NP_001153872.1 | 41 | 8790 | ||
GenBank mRNA | AK030408.1 | - | 14 | 3515 | ||
AK122377.1 | - | 18 | 4278 | |||
BC031185.1 | - | 1 | 828 | |||
BC036727.1 | - | 18 | 4432 | |||
BC060277.1 | - | 17 | 3970 |