Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micall1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177461.1 | NP_803412.1 | 16 | 4036 | ||
GenBank mRNA | AK076226.1 | - | 3 | 1337 | ||
AK129419.1 | - | 16 | 3775 | |||
AK147587.1 | - | 16 | 3837 | |||
AK147646.1 | - | 16 | 4049 |