Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290729.1 | NP_001277658.1 | 38 | 5473 | ||
GenBank mRNA | AB299229.1 | - | 38 | 5400 | ||
AK046668.1 | - | 16 | 1956 | |||
AK132088.1 | - | 3 | 335 | |||
BC047418.1 | - | 27 | 3580 | |||
BC094568.1 | - | 19 | 2751 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK049826.1 | - | 329 | Chr. - - [-] on Build |