Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290729.1 | NP_001277658.1 | 16 | 2635 | ||
GenBank mRNA | AB299229.1 | - | 16 | 2562 | ||
AF081115.1 | - | 1 | 30 | |||
BC047418.1 | - | 6 | 748 | |||
BC051073.1 | - | 3 | 361 |