Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012723.2 | NP_001012741.2 | 4 | 719 | ||
GenBank mRNA | BC043944.1 | - | 4 | 705 |