Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012723.2 | NP_001012741.2 | 2, 3 | 217 | ||
GenBank mRNA | AY542488.1 | - | 2, 3 | 217 | ||
AY542489.1 | - | 2, 3 | 217 | |||
BC043944.1 | - | 2, 3 | 211 | |||
BC145970.1 | - | 1, 2 | 164 | 40131273 | ||
BC145972.1 | - | 1, 2 | 164 | 40131275 |