Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup62cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081668.1 | NP_001075137.1 | 4 | 485 | ||
GenBank mRNA | BC100367.1 | - | 4 | 469 |