Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsperg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164658.1 | NP_001158130.1 | 28 | 3297 | ||
GenBank mRNA | AK133307.1 | - | 27 | 3304 | ||
BC172039.1 | - | 21 | 2278 |