Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dopey1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177208.3 | NP_796182.2 | 4 | 500 | ||
GenBank mRNA | AK044820.1 | - | 3 | 390 | ||
AK045873.1 | - | 3 | 387 | |||
AK052544.1 | - | 3 | 389 | |||
AK054511.1 | - | 3 | 431 | |||
AK082462.1 | - | 3 | 422 |