Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dopey1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177208.3 | NP_796182.2 | 28, 29 | 6153 | ||
GenBank mRNA | AK038064.1 | - | 5, 6 | 1229 | ||
AK047998.1 | - | 6, 7 | 658 | |||
AK169824.1 | - | 2, 3 | 108 | |||
BC043710.1 | - | 3, 4 | 374 |