Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpy19l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166207.1 | NP_001159679.1 | 9 | 1206 | ||
GenBank mRNA | AK029497.1 | - | 9 | 1209 | ||
AK030128.1 | - | 9 | 1196 |