Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081132.1 | NP_001074601.1 | 14 | 2915 | ||
GenBank mRNA | AK045943.1 | - | 1 | 561 | ||
BC039278.1 | - | 3 | 578 | |||
BC138358.1 | - | 14 | 2878 | 8861378 | ||
BC138359.1 | - | 14 | 2878 | 8861379 |