Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apol11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143686.1 | NP_001137158.1 | 4 | 1370 | ||
GenBank mRNA | AK039744.1 | - | 3 | 1249 | ||
AY662646.1 | - | 5 | 1605 |