Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apol11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143686.1 | NP_001137158.1 | 3 | 210 | ||
GenBank mRNA | BB653856.1 | - | 3 | 211 |