Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163319.1 | NP_001156791.1 | 25 | 5617 | ||
GenBank mRNA | AK052293.1 | - | 2 | 580 | ||
BC023717.1 | - | 4 | 546 | |||
BC057626.1 | - | 10 | 2524 |