Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AI464131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085515.2 | NP_001078984.1 | 2 | 3825 | ||
GenBank mRNA | AK173117.1 | - | 2 | 3809 | ||
BC036141.1 | - | 1 | 2577 |