Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_184088.1 | NP_908976.1 | 25 | 4911 | ||
GenBank mRNA | AK155029.1 | - | 4 | 714 | ||
BC040249.1 | - | 10 | 2318 | |||
BC040798.1 | - | 10 | 2318 | |||
BC146602.1 | - | 24 | 4536 | 40099465 |