Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_184088.1 | NP_908976.1 | 13 | 2275 | ||
GenBank mRNA | AK155220.1 | - | 13 | 2276 | ||
AK158927.1 | - | 3 | 1721 | |||
BC146602.1 | - | 12 | 1900 | 40099465 |