Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017359.2 | NP_059055.2 | 2 | 219 | ||
GenBank mRNA | AY724493.1 | - | 2 | 398 | ||
BC085744.1 | - | 2 | 219 | 7191921 | ||
M83677.1 | - | 2 | 371 |