Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033540.2 | NP_001028712.1 | 3 | 2906 | ||
GenBank mRNA | AK142560.1 | - | 3 | 2693 | ||
AK147880.1 | - | 3 | 2907 |