Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033431.1 | NP_001028603.1 | 3 | 1343 | ||
GenBank mRNA | AK158158.1 | - | 3 | 1344 | ||
AY364010.1 | - | 2 | 1079 | |||
BC116252.1 | - | 3 | 1216 | 40093617 | ||
BC116253.1 | - | 3 | 1216 | 40093618 |