Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270617.1 | NP_001257546.1 | 2 | 412 | ||
NM_001270618.1 | NP_001257547.1 | 2 | 412 | |||
XM_008762140.1 | XP_008760362.1 | 2 | 484 | |||
XM_008762141.1 | XP_008760363.1 | 2 | 483 | |||
XM_008762142.1 | XP_008760364.1 | 2 | 484 | |||
XM_008762143.1 | XP_008760365.1 | 2 | 484 | |||
XM_008762144.1 | XP_008760366.1 | 2 | 484 | |||
GenBank mRNA | BC087017.1 | - | 2 | 454 | 7109809 | |
EF577032.1 | - | 1 | 396 | |||
FM059245.1 | - | 1 | 44 | |||
JN579708.1 | - | 2 | 412 | |||
U10096.1 | - | 2 | 412 |