Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc47a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033542.2 | NP_001028714.1 | 9, 10 | 972 | ||
GenBank mRNA | AK133432.1 | - | 9, 10 | 973 | ||
BC147619.1 | - | 9, 10 | 920 | 9056609 | ||
BC157913.1 | - | 9, 10 | 920 | 9088130 |