Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081336.1 | NP_001074805.1 | 9, 10 | 1117 | ||
NM_001253766.1 | NP_001240695.1 | 7, 8 | 805 | |||
NM_001281794.1 | NP_001268723.1 | 7, 8 | 805 | |||
GenBank mRNA | AK031069.1 | - | 7, 8 | 806 | ||
AK083819.1 | - | 7, 8 | 806 | |||
AK133314.1 | - | 7, 8 | 1545 | |||
BC117904.1 | - | 7, 8 | 789 | |||
BC127173.1 | - | 7, 8 | 789 | 40097093 |