Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099664.1 | NP_001093134.1 | 1 | 297 | ||
GenBank mRNA | AK131881.1 | - | 1 | 79 |