Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC010546.1 | - | 582 | 4018506 | ||
CT010351.1 | - | 496 | ||||
M31419.1 | - | 665 |
Gene Symbol: Mnda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033450.4 | NP_001028622.1 | 4 | 728 | ||
NM_001301745.1 | NP_001288674.1 | 4 | 722 | |||
GenBank mRNA | AK152278.1 | - | 4 | 596 | ||
AK152621.1 | - | 3 | 262 | |||
AK152683.1 | - | 4 | 596 | |||
AK152786.1 | - | 4 | 605 | |||
AK166217.1 | - | 4 | 728 | |||
BC132314.1 | - | 4 | 549 | 40130591 | ||
BC132316.1 | - | 4 | 549 | 40130593 | ||
BC145397.1 | - | 4 | 543 | 9053955 | ||
M74123.1 | - | 4 | 583 | |||
S62227.1 | - | 4 | 583 |
Gene Symbol: Mndal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FJ594741.1 | - | 767 |