Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034875.3 | NP_001030047.2 | 12 | 1642 | ||
GenBank mRNA | AK143420.1 | - | 9 | 1006 | ||
BC147610.1 | - | 9 | 1005 | 9056600 | ||
BC147625.1 | - | 9 | 1005 | 9056615 |