Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114079.2 | NP_001107551.1 | 2 | 464 | ||
GenBank mRNA | BC158030.1 | - | 2 | 464 | 9007333 | |
EU669908.1 | - | 2 | 207 |