Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctxn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162934.1 | NP_001156406.2 | 2 | 997 | ||
GenBank mRNA | AK141570.1 | - | 2 | 999 |