Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033453.3 | NP_001028625.1 | 2 | 3258 | ||
NM_001098230.1 | NP_001091700.1 | 3 | 3293 | |||
NM_001098231.1 | NP_001091701.1 | 2 | 3209 | |||
NM_001290387.1 | NP_001277316.1 | 2 | 3207 | |||
NM_001290391.1 | NP_001277320.1 | 3 | 3242 | |||
GenBank mRNA | AK133542.1 | - | 2 | 3201 |