Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpar5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163268.1 | NP_001156740.1 | 2 | 1356 | ||
NM_001163269.1 | NP_001156741.1 | 2 | 1291 | |||
GenBank mRNA | BC116702.1 | - | 2 | 1256 | 40104865 | |
BC117528.1 | - | 2 | 1256 | 40104864 |
Set Membership: |
G-protein coupled receptor |